Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
A suggested positive control is HepG2 cell lysate.
PA5-21074 can be used with blocking peptide PEP-1188.
The zinc transporter ZIP9, also known as SLC39A9, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP9 is a multipass membrane protein that belongs to the ZIP transporter subfamily I. It is located at the trans-Golgi network regardless of zinc presence and is thought to be a zinc homeostasis regulator acting in the secretory pathway without significantly altering cytosolic zinc homeostasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: solute carrier family 39 (zinc transporter), member 9; Solute carrier family 39 member 9; solute carrier family 39, member 9; zinc transporter SLC39A9; Zinc transporter ZIP9; ZIP-9; Zrt- and Irt-like protein 9
Gene Aliases: 2010002A02Rik; 2610511I23Rik; 4833420E20Rik; AW209151; SLC39A9; UNQ714/PRO1377; ZIP-9; ZIP9
UniProt ID: (Human) Q9NUM3, (Mouse) Q8BFU1
Entrez Gene ID: (Human) 55334, (Mouse) 328133
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support