Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAEDKSKRDS IEMSMKGCQT NNGFVHNEDI LEQTPDPGSS TDNLKHSTRG ILGSQEPDFK GVQPYAGMPK EVLFQFSGQA RYRIPREILF WLTVASVLVL IAATIAIIAL SPKCLDWWQE GPMYQIYPRS FKDSNKDGNG DLKGIQDKLD YITALNIKTV WITSFYKSSL KDFRYGVEDF REVDPIFGTM EDFENLVAAI HDKGLKLIID FIPNHTSDKH IWFQLSRTRT GKYTDYYIWH DCTHENGKTI PPNNWLSVYG NSSWHFDEVR NQCYFHQFMK EQPDLNFRNP DVQEEIKEIL RFWLTKGVDG FSLDAVKFLL EAKHLRDEIQ VNKTQIPDTV TQYSELYHDF TTTQVGMHDI VRSFRQTMDQ YSTEPGRYRF MGTEAYAESI DRTVMYYGLP FIQEADFPFN NYLSMLDTVS GNSVYEVITS WMENMPEGKW PNWMIGGPDS SRLTSRLGNQ YVNVMNMLLF TLPGTPITYY GEEIGMGNIV AANLNESYDI NTLRSKSPMQ WDNSSNAGFS EASNTWLPTN SDYHTVNVDV QKTQPRSALK LYQDLSLLHA NELLLNRGWF CHLRNDSHYV VYTRELDGID RIFIVVLNFG ESTLLNLHNM ISGLPAKIRI RLSTNSADKG SKVDTSGIFL DKGEGLIFEH NTKNLLHRQT AFRDRCFVSN RACYSSVLNI LYTSC
This gene encodes a type II membrane glycoprotein which is one of the components of the renal amino acid transporter which transports neutral and basic amino acids in the renal tubule and intestinal tract.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amino acid transporter 1; Amino acid transporter heavy chain SLC3A1; B(0,+)-type amino acid transport protein; b(0,+)-type amino acid transporter-related heavy chain; D2h; FLJ34681; NBAT; Neutral and basic amino acid transport protein; rBAT; SLC3A1 variant B; SLC3A1 variant C; SLC3A1 variant D; SLC3A1 variant E; SLC3A1 variant F; SLC3A1 variant G; solute carrier family 3 (amino acid transporter heavy chain), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters, activator of cystine, dibasic and neutral amino acid transport), member 1; Solute carrier family 3 member 1
Gene Aliases: ATR1; CSNU1; D2H; NBAT; RBAT; SLC3A1
UniProt ID: (Human) Q07837
Entrez Gene ID: (Human) 6519
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support