Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MDTFSHAVSL LNFGPALATT QRVRDCCCGV SLVCPSASHQ HAPLLRDTSS LPPSLVPQAC REGPLLPRAP GGVLPFTTWE RCQFSSELNK ARAHSMLGAQ PKVLVTSSCK ASHHPPARAQ GGPLASPSLG PPGGLSTPPS GIPCPPQCCQ GHVALCRGLR PSPGDRMTRL LEMPRCQRNS PGISERNYLV PL
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Solute carrier family 45 member 4; solute carrier family 45, member 4
Gene Aliases: KIAA1126; SLC45A4
UniProt ID: (Human) Q5BKX6
Entrez Gene ID: (Human) 57210
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support