Search Thermo Fisher Scientific
图: 1 / 3
Immunogen sequence: QAVCFLGFI IQLNWKKACQ QAQVHANLKV NNVPRSGNSA LPQDPLHPGC PENLEGILTN DVGKTGEPQS DQQMRQEEPL PEHPQDGAKL SRKQLVLRRG LLLLGVFLIL LVGILVRFYV RIQ (449-570 aa encoded by BC010661)
This gene is located within the Smith-Magenis syndrome region on chromosome 17. It encodes a protein of unknown function.
仅用于科研。不用于诊断过程。未经明确授权不得转售。
蛋白别名: FLJ10847; H+/organic cation antiporter variant 1; H+/organic cation antiporter variant 2; hMATE-1; MATE-1; MGC64822; mMATE-1; multidrug and toxin extrusion 1; Multidrug and toxin extrusion protein 1; rMATE-1; solute carrier family 47 (multidrug and toxin extrusion), member 1; Solute carrier family 47 member 1; solute carrier family 47, member 1
基因别名: 1300013J15Rik; AI036982; MATE1; mMATE1; RGD1311123; SLC47A1
UniProt ID: (Human) Q96FL8, (Rat) Q5I0E9, (Mouse) Q8K0H1
Entrez Gene ID: (Human) 55244, (Rat) 360539, (Mouse) 67473