Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
This gene is located within the Smith-Magenis syndrome region on chromosome 17. It encodes a protein of unknown function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10847; H+/organic cation antiporter variant 1; H+/organic cation antiporter variant 2; MATE-1; MGC64822; mMATE-1; Multidrug and toxin extrusion protein 1; rMATE-1; Solute carrier family 47 member 1; solute carrier family 47, member 1
Gene Aliases: 1300013J15Rik; AI036982; MATE1; mMATE1; RGD1311123; Slc47a1
UniProt ID: (Rat) Q5I0E9, (Mouse) Q8K0H1
Entrez Gene ID: (Rat) 360539, (Mouse) 67473
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support