Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: DDYEDMMEEN LEQEEYEDPD IPESQMEEPA AHDTEATATD YHTTSHPGTH KVYVELQELV MDEKNQELRW MEAARWVQLE ENLGENGAWG RPHLSHLT
Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 58%.
The Band 3 protein is the most abundant integral protein of human erythrocyte membranes and functions as the major anion transporter polypeptide of erythrocytes. Band 3 is a 90-100 kDa protein, its microheterogeneity is ascribed mainly to the structural heterogeneity of the carbohydrate moiety. The C-terminal portion of the molecule spans the membrane several times, and is responsible for anion transport. The N-terminal portion, comprising almost half of the polypeptide chain of Band 3, is located inside the red cell and interacts with cytoskeletal and cytoplasmic proteins such as ankyrin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AE 1; Anion exchange protein 1; anion exchanger 1; anion exchanger-1; Band 3 anion transport protein; CAMP; CD233; Diego blood group; erythrocyte membrane protein band 3; erythroid anion exchange protein; Froese blood group; MGC116750; MGC116753; MGC126619; MGC126623; solute carrier family 4 (anion exchanger), member 1 (Diego blood group); Solute carrier family 4 member 1; solute carrier family 4, anion exchanger, member 1 (erythrocyte membrane protein band 3, Diego blood group); solute carrier family 4, anion exchanger, number 1; Swann blood group; Waldner blood group; Wright blood group; ZNF828
Gene Aliases: AE1; BND3; CD233; CHC; DI; EMPB3; EPB3; FR; RTA1A; SAO; SLC4A1; SPH4; SW; WD; WD1; WR
UniProt ID: (Human) P02730
Entrez Gene ID: (Human) 6521
Molecular Function: secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support