Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MSTENVEGKP SNLGERGRAR SSTFLRVVQP MFNHSIFTSA VSPAAERIRF ILGEEDDSPA PPQLFTELDE LLAVDGQEME WKETARWIKF EEKVEQGGER WSKPHVATLS LHSLFELRTC MEKGSIMLDR EASSLPQLVE MIVDHQIETG LLKPELKDKV TYTLLRKHRH QTKKSNLRSL ADIGKTVSSA SRMFTNPDNG SPAMTHRNLT SSSLNDISDK PEKDQLKNKF MKKLPRDAEA SNVLVGEVDF LDTPFIAFVR LQQAVMLGAL TEVPVPTRFL FILLGPKGKA KSYHEIGRAI ATLMSDEVFH DIAYKAKDRH DLIAGIDEFL DEVIVLPPGE WDPAIRIEPP KSLPSSDKRK NMYSGGENVQ MNGDTPHDGG HGGGGHGDCE ELQRTGRFCG GLIKDIKRKA PFFASDFYDA LNIQALSAIL FIYLATVTNA ITFGGLLGDA TDNMQGVLES FLGTAVSGAI FCLFAGQPLT ILSSTGPVLV FERLLFNFSK DNNFDYLEFR LWIGLWSAFL CLILVATDAS FLVQYFTRFT EEGFSSLISF IFIYDAFKKM IKLADYYPIN SNFKVGYNTL FSCTCVPPDP GEGITLCVYA RFVFGGRCRL HACKFSTCCH GPQELVLFFS LKNSATEFDV SLPEVF
Slc4a4 encodes a sodium bicarbonate cotransporter (NBC) involved in the regulation of bicarbonate secretion and absorption and intracellular pH. Slc4a4 is an electrogenic sodium/bicarbonate cotransporter with a Na(+):HCO3(-) stoichiometry varying from 1:2 to 1:3. Moreover, Slc4a4 may regulate bicarbonate influx/efflux at the basolateral membrane of cells and regulate intracellular pH. Isoform 2: May have a higher activity than isoform 1. Mutations in the Slc4a4 gene are associated with proximal renal tubular acidosis. Multiple transcript variants encoding different isoforms have been found for the Slc4a4 gene. Diseases associated with SLC4A4 include Renal Tubular Acidosis, Proximal, With Ocular Abnormalities and Proximal Renal Tubular Acidosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Electrogenic sodium bicarbonate cotransporter 1; HNBC 1; kNBC 1; kNBC1; Na(+)/HCO3(-) cotransporter; NBC 1; NBC 2; NBCE 1; OTTHUMP00000160355; OTTHUMP00000218884; OTTHUMP00000218885; Sodium bicarbonate cotransporter; sodium bicarbonate cotransporter 1 (sodium bicarbonate cotransporter, kidney; sodium bicarbonate cotransporter, pancreas); solute carrier family 4 (sodium bicarbonate cotransporter), member 4; Solute carrier family 4 member 4; solute carrier family 4, sodium bicarbonate cotransporter, member 4, brain type; solute carrier family 4, sodium bicarbonate cotransporter, member 5
Gene Aliases: hhNMC; HNBC1; KNBC; kNBC1; NBC; NBC1; NBC2; NBCE1; NBCe1-A; pNBC; SLC4A4; SLC4A5
UniProt ID: (Human) Q9Y6R1
Entrez Gene ID: (Human) 8671
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support