Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 7
Antibody detects endogenous levels of total SLC4A7.
SLC4A7 regulates intracellular pH and may also play a role in bicarbonate salvage in secretory epithelia. Additionally, SLC4A7 may also have associated sodium channel activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bicarbonate transporter; Electroneutral Na/HCO(3) cotransporter; Electroneutral sodium bicarbonate cotransporter 1; Na-bicarbonate cotransporter; NBC-like protein; Sodium bicarbonate cotransporter 2; Sodium bicarbonate cotransporter 2b; Sodium bicarbonate cotransporter 3; sodium bicarbonate cotransporter NBCn1-A; sodium bicarbonate cotransporter NBCn1-B; sodium bicarbonate cotransporter NBCn1-c; sodium bicarbonate cotransporter NBCn1-D; sodium bicarbonate cotransporter NBCn1-H; Solute carrier family 4 member 7; solute carrier family 4, sodium bicarbonate cotransporter, member 7
Gene Aliases: BT; E430014N10Rik; Nbc; NBC2; NBC2B; NBC3; NBCN1; NBCn1-G; SBC2; SLC4A6; SLC4A7
UniProt ID: (Human) Q9Y6M7, (Rat) Q9R1L1
Entrez Gene ID: (Human) 9497, (Rat) 117955, (Mouse) 218756
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support