Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total SLC4A7 protein.
SLC4A7 regulates intracellular pH and may also play a role in bicarbonate salvage in secretory epithelia. Additionally, SLC4A7 may also have associated sodium channel activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bicarbonate transporter; Electroneutral Na/HCO(3) cotransporter; Sodium bicarbonate cotransporter 2; Sodium bicarbonate cotransporter 2b; Sodium bicarbonate cotransporter 3; Solute carrier family 4 member 7; solute carrier family 4, sodium bicarbonate cotransporter, member 7
Gene Aliases: BT; NBC2; NBC2B; NBC3; NBCN1; SBC2; SLC4A6; SLC4A7
UniProt ID: (Human) Q9Y6M7
Entrez Gene ID: (Human) 9497
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support