Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: IMR32, U87-MG.
Predicted reactivity: Mouse (92%), Rat (91%), Rhesus Monkey (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Mediates electroneutral sodium- and carbonate-dependent choride-HCO3(-) exchange with a Na(+):HCO3(-) stoichiometry of 2:1. Plays a major role in pH regulation in neurons. May be involved in cell pH regulation by transporting HCO3(-) from blood to cell. Enhanced expression in severe acid stress could be important for cell survival by mediating the influx of HCO3(-) into the cells. Also mediates lithium-dependent HCO3(-) cotransport. May be regulated by osmolarity.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Electroneutral Na(+)-driven Cl-HCO3 exchanger; Electroneutral sodium bicarbonate exchanger 1; k-NBC3; Solute carrier family 4 member 8; solute carrier family 4, sodium bicarbonate cotransporter, member 8
Gene Aliases: KIAA0739; NBC; NBC3; NDCBE; NDCBE1; SLC4A8
UniProt ID: (Human) Q2Y0W8
Entrez Gene ID: (Human) 9498
Molecular Function: secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support