Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: IEEISDLILD QQELSSDLND SMRVKVREAL LKKHHHQNEK KRNNLIPIVR SFAEVGKKQS DPHLMDKHGQ TVSPQSVPTT NLEVKNGVNC EHSPVDLSKV
Mediates electroneutral sodium- and carbonate-dependent choride-HCO3(-) exchange with a Na(+):HCO3(-) stoichiometry of 2:1. Plays a major role in pH regulation in neurons. May be involved in cell pH regulation by transporting HCO3(-) from blood to cell. Enhanced expression in severe acid stress could be important for cell survival by mediating the influx of HCO3(-) into the cells. Also mediates lithium-dependent HCO3(-) cotransport. May be regulated by osmolarity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Electroneutral Na(+)-driven Cl-HCO3 exchanger; Electroneutral Na+-driven Cl-HCO3 exchanger; Electroneutral sodium bicarbonate exchanger 1; k-NBC3; slc4a8; sodium bicarbonate cotransporter isoform 3 kNBC-3; Solute carrier family 4 member 8; solute carrier family 4, sodium bicarbonate cotransporter, member 8
Gene Aliases: AW493845; C230026C11; KIAA0739; kNBC-3; NBC; NBC3; NDCBE; NDCBE1; SLC4A8
UniProt ID: (Human) Q2Y0W8, (Mouse) Q8JZR6
Entrez Gene ID: (Human) 9498, (Mouse) 59033
Molecular Function: secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support