Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RMRGRSLNP ATIYPVLPKL LSLLPLSCQK RLHCRSYGQD HLDTGLFPEK PRNGVLGDSR DKEAMALDGT AYQGSSSTCI LQETSL (551-635 aa encoded by BC012806)
Transports pantothenate, biotin and lipoate in the presence of sodium. Target protein mediates sodium-dependent transport of pantothenate, biotin, and lipoate; plays a role in intestinal uptake of biotin and in transplacental vitamin transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Na(+)-dependent multivitamin transporter; Na+-dependent multivitamin transporter; Sodium-dependent multivitamin transporter; solute carrier family 5 (sodium-dependent vitamin transporter), member 6; solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6; Solute carrier family 5 member 6; solute carrier family 5, member 6
Gene Aliases: E430023I20; SLC5A6; SMVT
UniProt ID: (Human) Q9Y289, (Mouse) Q5U4D8, (Rat) O70247
Entrez Gene ID: (Human) 8884, (Mouse) 330064, (Rat) 170551
Molecular Function: secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support