Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Transports betaine and GABA. May have a role in regulation of GABAergic transmission in the brain through the reuptake of GABA into presynaptic terminals, as well as in osmotic regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Betaine/GABA transporter 1; betaine/GABA transporter-1; BGT-1; GABA transporter 1; gamma-aminobutyric acid (GABA-A) transporter 2; gamma-aminobutyric acid transporter; GAT-2; Na(+)/Cl(-) betaine/GABA transporter; Sodium- and chloride-dependent betaine transporter; sodium- and chloride-dependent GABA transporter 2; solute carrier family 6 (neurotransmitter transporter), member 12; solute carrier family 6 (neurotransmitter transporter, betaine/GABA), member 12; Solute carrier family 6 member 12; vesicular GABA transporter
Gene Aliases: BGT-1; BGT1; Gabt2; Gat1; GAT2; RNU28927; SLC6A12; VGAT
UniProt ID: (Human) P48065, (Rat) P48056
Entrez Gene ID: (Human) 6539, (Mouse) 14411, (Rat) 50676
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support