Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
This gene encodes a member of the solute carrier family 6. Members of this family are sodium and chloride dependent neurotransmitter transporters. The encoded protein transports both neutral and cationic amino acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amino acid transporter ATB0+; amino acid transporter B0+; ATB(0+); colonic system B0+ amino acid transporter; Colonic system B0+ amino acid transporter CATB0+; neutral cationic amino acid transporter B0,+; Sodium- and chloride-dependent neutral and basic amino acid transporter B(0+); solute carrier family 6 (amino acid transporter), member 14; solute carrier family 6 (neurotransmitter transporter), member 14; Solute carrier family 6 member 14
Gene Aliases: 1110007A17Rik; 9030613J17Rik; Atb0+; ATB0plus; BMIQ11; CATB0plus; SLC6A14
UniProt ID: (Human) Q9UN76, (Mouse) Q9JMA9
Entrez Gene ID: (Human) 11254, (Mouse) 56774, (Rat) 298340
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support