Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total SLC6A15.
SLC6A15 (solute carrier family 6 (neutral amino acid transporter), member 15), also known as sodium-dependent neutral amino acid transporter B(0)AT2, transporter v7-3, NTT73 or sodium-coupled branched-chain amino-acid transporter 1 (SBAT1), is a 730 amino acid multi-pass membrane protein that acts as a sodium-dependent neutral amino acid transporter. A member of the sodium neurotransmitter symporter (SNF) family and SLC6A15 subfamily, SLC6A15 differs from other members of its family in that it does not appear to be chloride-dependent. SLC6A15 is expressed in brain and is encoded by a gene that maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4. 5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and trisomy 12p, which causes facial developmental defects and seizure disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homolog of rat orphan transporter v7-3; orphan sodium- and chloride-dependent neurotransmitter transporter NTT73; orphan transporter v7-3; Sodium- and chloride-dependent neurotransmitter transporter NTT73; Sodium-coupled branched-chain amino-acid transporter 1; Sodium-dependent neutral amino acid transporter B(0)AT2; sodium/chloride dependent neurotransmitter transporter Homo sapiens orphan neurotransmitter transporter NTT7; solute carrier family 6 (neurotransmitter transporter), member 15; solute carrier family 6 (neutral amino acid transporter), member 15; Solute carrier family 6 member 15; solute carrier family 6, member 15; Transporter v7-3
Gene Aliases: B0AT2; hv7-3; NTT73; SBAT1; SLC6A15; V7-3
UniProt ID: (Human) Q9H2J7
Entrez Gene ID: (Human) 55117
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support