Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: RCCERNAEIL LKLINLGKLP PDAKPPVNLL YNPTSIYNAW LSGLPQHIKS MVLREVTECN IETQFLKASE GPKFAFLSFV EAMSFLP
SLC6A16 shows structural characteristics of an Na(+)- and Cl(-)-dependent neurotransmitter transporter, including 12 transmembrane (TM) domains, intracellular N and C termini, and large extracellular loops containing multiple N-glycosylation sites.[supplied by OMIM, Mar 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTC-301O7.4; FLUP; FREQ; Orphan sodium- and chloride-dependent neurotransmitter transporter NTT5; solute carrier family 6 (neurotransmitter transporter), member 16; Solute carrier family 6 member 16; solute carrier family 6, member 16
Gene Aliases: NT5; NTT5; SLC6A16
UniProt ID: (Human) Q9GZN6
Entrez Gene ID: (Human) 28968
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support