Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: METTPLNSQK QLSACEDGED CQENGVLQKV VPTPGDKVES GQISNGYSAV PSPGAGDDTR HSIPATTTTL VAELHQGERE TWGKKVDFLL SVIGYAVDLG NVWRFPYICY QNGGGAFLLP YTIMAIFGGI PLFYMELALG QYHRNGCISI WRKICPIFKG IGYAICIIAF YIASYYNTIM AWALYYLISS FTDQLPWTSC KNSWNTGNCT NYFSEDNITW TLHSTSPAEE FYTRHVLQIH RSKGLQDLGG ISWQLALCIM LIFTVIYFSI WKGVKTSGKV VWVTATFPYI ILSVLLVRGA TLPGAWRGVL FYLKPNWQKL LETGVWIDAA AQIFFSLGPG FGVLLAFASY NKFNNNCYQD ALVTSVVNCM TSFVSGFVIF TVLGYMAEMR NEDVSEVAKD AGPSLLFITY AEAIANMPAS TFFAIIFFLM LITLGLDSTF AGLEGVITAV LDEFPHVWAK RRERFVLAVV ITCFFGSLVT LTFGGAYVVK LLEEYATGPA VLTVALIEAV AVSWFYGITQ FCRDVKEMLG FSPGWFWRIC WVAISPLFLL FIICSFLMSP PQLRLFQYNY PYWSIILGYC IGTSSFICIP TYIAYRLIIT PGTFKERIIK SITPETPTEI PCGDIRLNAV
SERT is an integral membrane protein that transports the neurotransmitter serotonin from synaptic spaces into presynaptic neurons. The encoded protein terminates the action of serotonin and recycles it in a sodium-dependent manner. This protein is a target of psychomotor stimulants, such as amphetamines and cocaine, and is a member of the sodium:neurotransmitter symporter family. A repeat length polymorphism in the promoter of this gene has been shown to affect the rate of serotonin uptake and may play a role in sudden infant death syndrome, aggressive behavior in Alzheimer disease patients, and depression-susceptibility in people experiencing emotional trauma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5 OCD1; 5-hydroxytryptamine (serotonin) transporter; 5HT transporter; 5HTT; HGNC:11050; Na+/Cl- dependent serotonin transporter; OTTHUMP00000163633; serotonin transporter 1; SERT; Sodium-dependent serotonin transporter; Sodium-dependent serotonin transporter (5HT transporter) (5HTT); solute carrier family 6 (neurotransmitter transporter), member 4; solute carrier family 6 (neurotransmitter transporter, serotonin), member 4; Solute carrier family 6 member 4
Gene Aliases: 5-HTT; 5-HTTLPR; 5HTT; AI323329; hSERT; HTT; OCD1; SERT; SERT1; SLC6A4
UniProt ID: (Human) P31645, (Mouse) Q60857
Entrez Gene ID: (Human) 6532, (Mouse) 15567
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support