Product References
Cottonseed oil alleviates ischemic stroke injury by inhibiting ferroptosis.
Brain and behavior
Sun M,Liu M,Li Q,Zhang X,Liu S,Yang H,Yang L,Tian J,Mi W,Ma Y
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Sun Oct 01 00:00:00 EDT 2023
miR-16-5p Regulates Ferroptosis by Targeting SLC7A11 in Adriamycin-Induced Ferroptosis in Cardiomyocytes.
Journal of inflammation research
Chen Y,Deng Y,Chen L,Huang Z,Yan Y,Huang Z
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Tue Feb 20 00:00:00 EST 2024
Slc7a11 stimulates glutathione synthesis to preserve fatty acid metabolism in primary hepatocytes.
Redox report : communications in free radical research
Liu Y,Wu K,Fu Y,Li W,Zhao XY
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Fri Dec 01 00:00:00 EST 2023
Transcriptome Profiles of IncRNA and mRNA Highlight the Role of Ferroptosis in Chronic Neuropathic Pain With Memory Impairment.
Frontiers in cell and developmental biology
Tang Y,Liu C,Zhu T,Chen H,Sun Y,Zhang X,Zhao Q,Wu J,Fei X,Ye S,Chen C
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Sat Jul 16 00:00:00 EDT 2022
TRIM26 Induces Ferroptosis to Inhibit Hepatic Stellate Cell Activation and Mitigate Liver Fibrosis Through Mediating SLC7A11 Ubiquitination.
Frontiers in cell and developmental biology
Zhu Y,Zhang C,Huang M,Lin J,Fan X,Ni T
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Immunoprecipitation
Wed Apr 21 00:00:00 EDT 2021
TRIM26 Induces Ferroptosis to Inhibit Hepatic Stellate Cell Activation and Mitigate Liver Fibrosis Through Mediating SLC7A11 Ubiquitination.
Frontiers in cell and developmental biology
Zhu Y,Zhang C,Huang M,Lin J,Fan X,Ni T
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Immunoprecipitation
Wed Apr 21 00:00:00 EDT 2021
Overexpression of Cystine/Glutamate Antiporter xCT Correlates with Nutrient Flexibility and ZEB1 Expression in Highly Clonogenic Glioblastoma Stem-like Cells (GSCs).
Cancers
Koch K,Hartmann R,Suwala AK,Rios DH,Kamp MA,Sabel M,Steiger HJ,Willbold D,Sharma A,Kahlert UD,Maciaczyk J
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Mon Nov 29 00:00:00 EST 2021
Overexpression of Cystine/Glutamate Antiporter xCT Correlates with Nutrient Flexibility and ZEB1 Expression in Highly Clonogenic Glioblastoma Stem-like Cells (GSCs).
Cancers
Koch K,Hartmann R,Suwala AK,Rios DH,Kamp MA,Sabel M,Steiger HJ,Willbold D,Sharma A,Kahlert UD,Maciaczyk J
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Mon Nov 29 00:00:00 EST 2021
Mitochondrial fusion supports increased oxidative phosphorylation during cell proliferation.
eLife
Yao CH,Wang R,Wang Y,Kung CP,Weber JD,Patti GJ
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Tue Jan 29 00:00:00 EST 2019
Mitochondrial fusion supports increased oxidative phosphorylation during cell proliferation.
eLife
Yao CH,Wang R,Wang Y,Kung CP,Weber JD,Patti GJ
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Tue Jan 29 00:00:00 EST 2019
Glutamate Signaling in Hepatic Stellate Cells Drives Alcoholic Steatosis.
Cell metabolism
Choi WM,Kim HH,Kim MH,Cinar R,Yi HS,Eun HS,Kim SH,Choi YJ,Lee YS,Kim SY,Seo W,Lee JH,Shim YR,Kim YE,Yang K,Ryu T,Hwang JH,Lee CH,Choi HS,Gao B,Kim W,Kim SK,Kunos G,Jeong WI
PA1-16893 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot to investigate hepatocyte cannabinoid receptor-1 in alcoholic liver disease and lipogenesis.
Tue Nov 05 00:00:00 EST 2019
Participation of xCT in melanoma cell proliferation in vitro and tumorigenesis in vivo.
Oncogenesis
Shin SS,Jeong BS,Wall BA,Li J,Shan NL,Wen Y,Goydos JS,Chen S
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Wed Nov 14 00:00:00 EST 2018
Participation of xCT in melanoma cell proliferation in vitro and tumorigenesis in vivo.
Oncogenesis
Shin SS,Jeong BS,Wall BA,Li J,Shan NL,Wen Y,Goydos JS,Chen S
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Wed Nov 14 00:00:00 EST 2018
Participation of xCT in melanoma cell proliferation in vitro and tumorigenesis in vivo.
Oncogenesis
Shin SS,Jeong BS,Wall BA,Li J,Shan NL,Wen Y,Goydos JS,Chen S
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Wed Nov 14 00:00:00 EST 2018
Comparative analysis of antibodies to xCT (Slc7a11): Forewarned is forearmed.
The Journal of comparative neurology
Van Liefferinge J,Bentea E,Demuyser T,Albertini G,Follin-Arbelet V,Holmseth S,Merckx E,Sato H,Aerts JL,Smolders I,Arckens L,Danbolt NC,Massie A
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Fri Apr 01 00:00:00 EDT 2016
Comparative analysis of antibodies to xCT (Slc7a11): Forewarned is forearmed.
The Journal of comparative neurology
Van Liefferinge J,Bentea E,Demuyser T,Albertini G,Follin-Arbelet V,Holmseth S,Merckx E,Sato H,Aerts JL,Smolders I,Arckens L,Danbolt NC,Massie A
Published figure using SLC7A11 polyclonal antibody (Product # PA1-16893) in Western Blot
Fri Apr 01 00:00:00 EDT 2016
Reduced expression of miRNA-27a modulates cisplatin resistance in bladder cancer by targeting the cystine/glutamate exchanger SLC7A11.
Clinical cancer research : an official journal of the American Association for Cancer Research
Drayton RM,Dudziec E,Peter S,Bertz S,Hartmann A,Bryant HE,Catto JW
PA1-16893 was used in immunohistochemistry to study the role of SLC7A11 in the mechanism by which miRNA-27a expression levels modulate bladder cancer cisplatin resistance
Tue Apr 01 00:00:00 EDT 2014