Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
SLC7A11 (also known as XCT) is a member of a hetromeric, sodium-independent, anionic amino acid transport system. It is highly specific for cysteine and glutamate. SLC7A11 is a mediator of Kaposi sarcoma-associated herpes virus fusion and entry into cells. Increased expression of this protein in primary gliomas is associated with neuronal cell death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amino acid transport system xc-; Calcium channel blocker resistance protein CCBR1; cysteine/glutamate transporter; Cystine/glutamate transporter; sodium independent anionic amino acid transport system; solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11; Solute carrier family 7 member 11; solute carrier family 7, (cationic amino acid transporter, y+ system) member 11; Subtle gray protein; xCT
Gene Aliases: 9930009M05Rik; AI451155; CCBR1; SLC7A11; sut; xCT
UniProt ID: (Human) A8K2U4, (Mouse) Q9WTR6
Entrez Gene ID: (Human) 23657, (Mouse) 26570
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support