Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QKSSPPSSP GPASPGPLTK QQSSFSDHLQ LVGTVHASVP EPGELKPALR PYLGFLDGYS PGAVVTWALG VMLASAITIG CVLVFGNSTL HLPHWGYILL LLLTSVMFLL SLLVLGAHQQ QYREDLFQIP MVPLIPALSI VLNICLMLKL SYLTWVRFSI WLLMGLAVYF GYGIRHSKEN QRELPGLNST HYVVFPRGSL EETVQAMQPP SQAPAQDPGH ME (415-635 aa encoded by BC008814)
Involved in the transport of the cationic amino acids (arginine, lysine and ornithine).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAT-4; CAT4; Cationic amino acid transporter 4; Ig heavy chain variable region; solute carrier family 7 (cationic amino acid transporter, y+ system), member 4; solute carrier family 7 (orphan transporter), member 4; Solute carrier family 7 member 4; solute carrier family 7, member 4; VH 3 family
Gene Aliases: AI853530; CAT-4; CAT4; HCAT3; SLC7A4; VH
UniProt ID: (Human) O43246, (Mouse) Q8BLQ7
Entrez Gene ID: (Human) 6545, (Mouse) 224022
Molecular Function:
amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support