Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Solute carrier family 7 member 5 also known as large neutral amino acids transporter small subunit 1 is a protein that in humans is encoded by the SLC7A5 gene.It is sodium-independent, high-affinity transport of large neutral amino acids such as phenylalanine, tyrosine, leucine, arginine and tryptophan, when associated with SLC3A2/4F2hc.It involved in cellular amino acid uptake and the transport of L-DOPA across the blood-brain barrier, and that of thyroid hormones triiodothyronine (T3) and thyroxine (T4) across the cell membrane in tissues such as placenta.Acts as an amino acid exchanger and plays a role in neuronal cell proliferation (neurogenesis) in brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4F2; 4F2 LC; 4F2 light chain; 4F2HC; 4F2LC; 4T2HC; CD98 light chain; CD98HC; E16; hLAT1; Integral membrane protein E16; L-type amino acid transporter 1; large neutral amino acids transporter 1; Large neutral amino acids transporter small subunit 1; LAT-1; LAT1; MDU1; MP hLAT1; NACAE; Protein TA1; sodium-independent neutral amino acid transporter LAT1; solute carrier family 7 (amino acid transporter light chain, L system), member 5; solute carrier family 7 (cationic amino acid transporter, y+ system), member 5; Solute carrier family 7 member 5; tumor-associated protein 1; y+ system cationic amino acid transporter
Gene Aliases: 4F2LC; CD98; CD98LC; D0H16S474E; D16S469E; E16; hLAT1; LAT1; MPE16; SLC7A5; TA1
UniProt ID: (Human) Q01650, (Mouse) Q9Z127, (Rat) Q63016
Entrez Gene ID: (Human) 8140, (Mouse) 20539, (Rat) 50719
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support