Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: SIGDAFSDFT LARQGYPRMA FSACFGGIIF NILVGVGLGC LLQISRSHTE
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
SLC24A6 belongs to a family of potassium-dependent sodium/calcium exchangers that maintain cellular calcium homeostasis through the electrogenic counter transport of 4 sodium ions for 1 calcium ion and 1 potassium ion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial sodium/calcium exchanger protein; Na(+)/K(+)/Ca(2+)-exchange protein 6; Sodium/calcium exchanger protein, mitochondrial; Sodium/potassium/calcium exchanger 6; solute carrier family 24 (sodium/lithium/calcium exchanger), member 6; solute carrier family 24 (sodium/potassium/calcium exchanger), member 6; Solute carrier family 24 member 6; solute carrier family 8 (sodium/lithium/calcium exchanger), member B1; Solute carrier family 8 member B1
Gene Aliases: NCKX6; NCLX; SLC24A6; SLC8B1
UniProt ID: (Human) Q6J4K2
Entrez Gene ID: (Human) 80024
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support