Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: SLC9A10.
SLC9A10 is a member of the sodium-hydrogen exchanger (NHE) family (see SLC9A1, MIM 107310) and is required for male fertility and sperm motility (Wang et al., 2003 [PubMed 14634667]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Na(+)/H(+) exchanger 10; NHE-10; sNHE; Sodium/hydrogen exchanger 10; Solute carrier family 9 member 10; Solute carrier family 9 member C1; solute carrier family 9, member 10; solute carrier family 9, subfamily C (Na+-transporting carboxylic acid decarboxylase), member 1; Sperm-specific Na(+)/H(+) exchanger; sperm-specific sodium proton exchanger
Gene Aliases: NHE; NHE-10; SLC9A10; SLC9C1; sperm-NHE
UniProt ID: (Human) Q4G0N8
Entrez Gene ID: (Human) 285335
Molecular Function:
ion channel
transporter
voltage-gated ion channel
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support