Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: TPMLTWLQI RVGVDLDENL KEDPSSQHQE ANNLDKNMTK AESARLFRMW YSFDHKYLKP ILTHSGPPLT TTLPEWCGPI SRLLTSPQAY GEQLKEDDVE CIVNQDELAI NYQEQASSPC SPPARLGLDQ KASPQTPGKE NIYEGDLGLG GYELKLEQTL GQSQLN (481-645 aa encoded by BC035779)
This gene encodes a sodium hydrogen exchanger that is a member of the solute carrier 9 protein family. The encoded protein localizes the to the late recycling endosomes and may play an important role in maintaining cation homeostasis. Defects in this gene are associated with attention-deficit/hyperactivity disorder.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ35613; Na(+)/H(+) exchanger 9; NHE-9; putative protein product of Nbla00118; Sodium/hydrogen exchanger; Sodium/hydrogen exchanger 9; sodium/proton exchanger NHE9; solute carrier family 9 (sodium/hydrogen exchanger), isoform 9; Solute carrier family 9 member 9; solute carrier family 9, subfamily A (NHE9, cation proton antiporter 9), member 9
Gene Aliases: 5730527A11Rik; 9930105B05; AI854429; AUTS16; Nbla00118; NHE9; RGD1560736; SLC9A9
UniProt ID: (Human) Q8IVB4, (Rat) D4A7H1, (Mouse) Q8BZ00
Entrez Gene ID: (Human) 285195, (Rat) 363115, (Mouse) 331004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support