Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Peptide sequence: DPQDVRPSVF HNIKLFVLCH SLLQLAQLMI SGYLKSSIST VEKRFGLSSQ
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
SLCO2B1 mediates the Na+-independent transport of organic anions such as taurocholate, the prostaglandins PGD2, PGE1, PGE2, leukotriene C4, thromboxane B2 and iloprost.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OATP-B; OATP-RP2; OATP2B1; Organic anion transporter B; Organic anion transporter polypeptide-related protein 2; Organic anion transporting polypeptide 2B1; solute carrier family 21 (organic anion transporter), member 9; Solute carrier family 21 member 9; Solute carrier organic anion transporter family member 2B1; solute carrier organic anion transporter family, member 2B1
Gene Aliases: KIAA0880; OATP-B; OATP2B1; OATPB; SLC21A9; SLCO2B1
UniProt ID: (Human) O94956
Entrez Gene ID: (Human) 11309
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support