Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Promo Code: RACCS25 Save on advanced culture solutions Save up to 22% on optimized formulations to streamline your neural, stem and cancer cell culture. Learn more
SLCO2B1 mediates the Na+-independent transport of organic anions such as taurocholate, the prostaglandins PGD2, PGE1, PGE2, leukotriene C4, thromboxane B2 and iloprost.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: multispecific organic anion transporter 1; OATP-B; OATP-RP2; OATP2B1; Organic anion transporter B; Organic anion transporter moatp1; Organic anion transporter polypeptide-related protein 2; Organic anion transporting polypeptide 2B1; solute carrier family 21 (organic anion transporter), member 9; Solute carrier family 21 member 9; Solute carrier organic anion transporter family member 2B1; solute carrier organic anion transporter family, member 2B1
Gene Aliases: AI060904; AI852653; KIAA0880; moat1; OATP-B; OATP2B1; OATPB; SLC21A9; SLCO2B1
UniProt ID: (Human) O94956, (Mouse) Q8BXB6, (Rat) Q9JHI3
Entrez Gene ID: (Human) 11309, (Mouse) 101488, (Rat) 140860
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support