Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: RNLYINNELQ DFTKTQMKPG VVPGCEPCRK LYCLHGICQP NATPGPMCHC EAGWVGLHCD QPADGPCHGH KCVHGQCVPL DALSYSCQCQ DGYSGALCNQ AGALAEPCRG LQCLHGHCQA SGTKGAHCVC DPGFSGELCE QESECRGDPV RDFHQVQRGY AICQTTRPLS WVECRGSCPG QGCCQGLRLK RRKFTFECSD GTSFAEEVEK PTKCGCALCA; Positive Samples: 293T
SLIT1 is thought to act as molecular guidance cue in cellular migration, and function appears to be mediated by interaction with roundabout homolog receptors.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Multiple EGF-like domains protein 4; Multiple epidermal growth factor-like domains protein 4; slit homolog 1; Slit homolog 1 protein; Slit-1
Gene Aliases: KIAA0813; MEGF4; SLIL1; SLIT-1; SLIT1; SLIT3
UniProt ID: (Human) O75093
Entrez Gene ID: (Human) 6585
Molecular Function:
immunoglobulin superfamily cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support