Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute at 0.2 mg/mL in sterile PBS.
Endoxin level is <0.20 EU per 1 µg of the antibody by the LAL method.
SLIT3 may act as molecular guidance cue in cellular migration, and function may be mediated by interaction with roundabout homolog receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Multiple EGF-like domains protein 5; multiple epidermal growth factor-like domains 5; Multiple epidermal growth factor-like domains protein 5; SLIL2FLJ10764; slit homolog 3; Slit homolog 3 protein; Slit-3
Gene Aliases: b2b2362.1Clo; KIAA0814; MEGF5; SLIL2; Slit-3; SLIT1; slit2; SLIT3; UNQ691/PRO1336
UniProt ID: (Human) O75094, (Rat) O88280, (Mouse) Q9WVB4
Entrez Gene ID: (Human) 6586, (Rat) 83467, (Mouse) 20564
Molecular Function:
immunoglobulin superfamily cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support