Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Keep as concentrated solution.
Positive Control: 293T, A549, H1299.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SLK acts as a novel effector of cytoskeletal remodeling and apoptosis, and seems to be ubiquitously expressed. SLK mediates apoptosis and actin stress fiber dissolution.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTCL tumor antigen se20-9; hSLK; Long Ste20-like Kinase; Serine/threonine-protein kinase 2; SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrose nonfermenting like kinase; SNF1 sucrose nonfermenting like kinase; STE20-like kinase; STE20-like serine/threonine-protein kinase; STE20-related kinase; Ste20-related serine/threonine kinase; STE20-related serine/threonine-protein kinase
Gene Aliases: bA16H23.1; KIAA0204; LOSK; se20-9; SLK; STK2
UniProt ID: (Human) Q9H2G2
Entrez Gene ID: (Human) 9748
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support