Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SLU7 Polyclonal Antibody detects endogenous levels of SLU7 protein.
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is a splicing factor that has been found to be essential during the second catalytic step in the pre-mRNA splicing process. It associates with the spliceosome and contains a zinc knuckle motif that is found in other splicing factors and is involved in protein-nucleic acid and protein-protein interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSlu7; MGC9280; Pre-mRNA-splicing factor SLU7; SLU7 splicing factor homolog; splicing factor; step II splicing factor SLU7; zinc knuckle motif containing
Gene Aliases: 9G8; AU018913; D11Ertd730e; D3Bwg0878e; hSlu7; SLU7
UniProt ID: (Human) O95391, (Mouse) Q8BHJ9, (Rat) Q80ZG5
Entrez Gene ID: (Human) 10569, (Mouse) 193116, (Rat) 303057
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support