Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Additional Information: This antibody is expected to recognise both reported isoforms (NP_003061.3; NP_620614.2).
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.
SMARCA2 encodes a protein that is a member of the SWI/SNF family of proteins and is highly similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. Two transcript variants encoding different isoforms have been found for this gene, which contains a trinucleotide repeat (CAG) length polymorphism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent helicase SMARCA2; BAF190B; BRG1-associated factor 190B; FLJ36757; global transcription activator homologous sequence; hBRM; MGC74511; Probable global transcription activator SNF2L2; Protein brahma homolog; SNF2-alpha; SNF2/SWI2-like protein 2; sucrose nonfermenting 2-like protein 2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 2
Gene Aliases: BAF190; BAF190B; BRM; hBRM; hSNF2a; NCBRS; SMARCA2; SNF2; SNF2A; SNF2L2; SNF2LA; Sth1p; SWI2
UniProt ID: (Human) P51531
Entrez Gene ID: (Human) 6595
Molecular Function:
DNA helicase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support