Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GLESSCIAG TGPDEPEKLE GAEEEKMEAD PDGQQPEKAE NKVENETDEG DKAQDGENEK NSEKEQDSEV SEDTKSEEKE TEENKELTDT CKERESDTGK KKVEHEISEG NVATAAAAAL ASAATKAKHL AAVEERKIKS LVALLVETQM KKLEIKLRHF EELETIMDRE KEALEQQRQQ LLTERQNFHM EQLKYAELRA RQQMEQQQHG QNPQQAHQHS GGPGLAPLGA AGHPGMMPHQ QPPPYPLMHH QMPPPHPPQP GQIPGPGSMM PGQHMPGRMI PTVAANIHPS GSGPTPPGMP PMPGNILGPR VPLTAPNGMY PPPPQQQPPP PPPADGVPPP PAPGPPASAA P (756-1105 aa encoded by BC050564)
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and contains a predicted leucine zipper motif typical of many transcription factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAF155; BRG1-associated factor 155; chromatin remodeling complex BAF155 subunit; mammalian chromatin remodeling complex BRG1-associated factor 155; SWI/SNF complex 155 kDa subunit; SWI/SNF complex subunit SMARCC1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 1; SWI3-related protein
Gene Aliases: AI115498; BAF155; CRACC1; Rsc8; SMARCC1; SRG3; SWI3
UniProt ID: (Human) Q92922, (Mouse) P97496
Entrez Gene ID: (Human) 6599, (Rat) 301020, (Mouse) 20588
Molecular Function: chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support