Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TLEDEAQEN KSKMKMVEEH MEQQKENMEH LKSLKIEAEN KYDAIKFKIN QLSELADPLK DELNLADSEV DNQKRGKRHY EEKQKEHLDT LNKKKRELDM KEKELEEKMS QARQICPERI EVEKSASILD KEINRLRQKI QAEHASHGDR EEIMRQYQEA RETYLDLDSK VRTLKKFIKL LGEIMEHRFK TYQQFRRCLT LRCKLYFDNL LSQRAYCGKM NFDHKNETLS ISVQPGEGNK AAFNDMRALS GGERSFSTVC FILSLWSIAE SPFRCLDEFD VYMDMVNRRI AMDLILKMAD SQRFRQFILL TPQSMSSLPS SKLIRILRMS DPERGQTTLP FRPVTQEEDD DQR (740-1091 aa encoded by B C039828)
Core component of the SMC5-SMC6 complex, a complex involved in DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintenance via recombination in ALT (alternative lengthening of telomeres) cell lines and mediates sumoylation of shelterin complex (telosome) components which is proposed to lead to shelterin complex disassembly in ALT-associated PML bodies (APBs). Required for recruitment of telomeres to PML nuclear bodies. SMC5-SMC6 complex may prevent transcription of episomal DNA, such as circular viral DNA genome (PubMed:26983541). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mSMC6; SMC protein 6; SMC6 structural maintenance of chromosomes 6-like 1; Structural maintenance of chromosomes protein 6
Gene Aliases: 2810489L22Rik; 3830418C19Rik; AA990493; AU018782; AW742439; hSMC6; Kiaa4103; mKIAA4103; SMC-6; SMC6; SMC6L1
UniProt ID: (Human) D6W518, (Mouse) Q924W5
Entrez Gene ID: (Human) 79677, (Rat) 313961, (Mouse) 67241
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support