Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 6
Immunogen sequence: DNGRGMTSKQL NNWAVYRLSK FTRQGDFESD HSGYVRPVPV PRSLNSDISY FGVGGKQAVF FVGQSARMIS KPADSQDVHE LVLSKED
Highest antigen sequence identity to the following orthologs - mouse 97%, rat 98%.
Required for maintenance of X inactivation in females and hypermethylation of CpG islands associated with inactive X. Involved in a pathway that mediates the methylation of a subset of CpG islands slowly and requires the de novo methyltransferase DNMT3B. Required for DUX4 silencing in somatic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SMC hinge domain-containing protein 1; Structural maintenance of chromosomes flexible hinge domain-containing protein 1
Gene Aliases: KIAA0650; SMCHD1
UniProt ID: (Human) A6NHR9
Entrez Gene ID: (Human) 23347
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support