Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Positive Samples: Mouse testis
Immunogen sequence: YKALESIEYV QTFKGLKTKY EQEKDRQNQK LNSVPSILRS NRFRRDAKAL EEDEEMWFNE DEEEEGKAVV APVEKPKPED DFPDNYEKFM ETKKAKESED KENLPKRTSP GGFKFTFSHS ASAANGTNSK SVVAQIPPAT SNGSSSKTTN LPTSVTATKG SLVGLVDYPD DEEEDEEEES SPRKRPRLGS
Regulatory subunit of serine/threonine-protein phosphatase 4 (PP4). May regulate the activity of PPP4C at centrosomal microtubule organizing centers. [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protein phosphatase 4, regulatory subunit 3B; Serine/threonine-protein phosphatase 4 regulatory subunit 3B; SMEK homolog 2; SMEK homolog 2, suppressor of mek1
Gene Aliases: AW011752; AW557776; FLFL2; KIAA1387; mKIAA1387; PP4R3B; PPP4R3B; PSY2; RGD1309450; SMEK2; smk1
UniProt ID: (Human) Q5MIZ7, (Mouse) Q922R5
Entrez Gene ID: (Human) 57223, (Mouse) 104570, (Rat) 360993
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support