Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
SMG5 plays a role in nonsense-mediated mRNA decay. SMG5 does not have RNase activity by itself. SMG5 promotes dephosphorylation of RENT1. SMG5 is necessary for TERT activity.SMG5 is involved in nonsense-mediated mRNA decay (Ohnishi et al., 2003 [PubMed 14636577]).[supplied by OMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EST1 telomerase component homolog B; EST1-like protein B; Est1p-like protein B; ever shorter telomeres 1B; FLJ34864; hSMG-5; LPTS interacting protein; LPTS-interacting protein; LPTS-RP1; Nonsense-mediated mRNA decay factor SMG5; RP11-54H19.7; SMG-5 homolog; smg-5 homolog, nonsense mediated mRNA decay factor; SMG5 nonsense mediated mRNA decay factor
Gene Aliases: EST1B; KIAA1089; LPTS-RP1; LPTSRP1; SMG-5; SMG5
UniProt ID: (Human) Q9UPR3
Entrez Gene ID: (Human) 23381
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support