Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Centrifuge briefly prior to opening.
This gene encodes a component of the telomerase ribonucleoprotein complex responsible for the replication and maintenance of chromosome ends. The encoded protein also plays a role in the nonsense-mediated mRNA decay (NMD) pathway, providing the endonuclease activity near the premature translation termination codon that is needed to initiate NMD. Alternatively spliced transcript variants encoding distinct protein isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EST1 telomerase component homolog A; Ever shorter telomeres 1A; hEST1A; hSmg5/7a; Nonsense mediated mRNA decay factor SMG6; Smg-6 homolog; smg-6 homolog, nonsense mediated mRNA decay factor; SMG6 nonsense mediated mRNA decay factor; telomerase subunit EST1A; Telomerase-binding protein EST1A
Gene Aliases: C17orf31; EST1A; hSMG5/7a; KIAA0732; SMG-6; SMG6
UniProt ID: (Human) Q86US8
Entrez Gene ID: (Human) 23293
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support