Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
SMG7 is involved in nonsense-mediated mRNA decay (Fukuhara et al., 2005 [PubMed 15721257]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: breast cancer-associated antigen SGA-56M; EST1 telomerase component homolog C; EST1-like protein C; ever shorter telomeres 1C; hSMG-7; Nonsense-mediated mRNA decay factor SMG7; protein SMG7; SMG-7 homolog; smg-7 homolog, nonsense mediated mRNA decay factor; SMG7 nonsense mediated mRNA decay factor
Gene Aliases: 9430023P16Rik; C1orf16; EST1C; KIAA0250; mKIAA0250; SGA56M; SMG7
UniProt ID: (Human) Q92540, (Mouse) Q5RJH6
Entrez Gene ID: (Human) 9887, (Mouse) 226517
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support