Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Antibody detects endogenous levels of total SMO.
Smoothened drosophila homolog (SMOH) is a serpentine protein belonging to the G-protein coupled receptor Fz/Smo family with 787 aa, seven transmembrane G protein-coupled receptors with 4 glycosylation sites and a putative extracellular amino terminus. Smoothened activated by b-Arrestin 2 and GRK2 acts as a receptor for Shh, the Sonic hedgehog gene product, and is a positive regulator of hedgehog signaling pathway along with the smoothened inhibitor patched protein (PTCH) and regulates vertebrate developmental processes, cellular proliferation and differentiation by activating transcription factor Gli. Smoothened mutants are known to cause sporadic basal cell carcinoma of the skin. It is known to express widely in heart and brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bent body; frizzled family member 11; HGNC:11119; Protein Gx; Protein SMOH; Protein smoothened; seven transmembrane helix receptor; smoothened homolog; smoothened, frizzled family receptor; smoothened, seven transmembrane spanning receptor
Gene Aliases: bnb; E130215L21Rik; FZD11; Gx; SMO; SMOH; smoothened
UniProt ID: (Human) Q99835, (Mouse) P56726, (Rat) P97698
Entrez Gene ID: (Human) 6608, (Mouse) 319757, (Rat) 25273
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support