Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MKPNFSLRLR IFNLNCWGIP YLSKHRADRM RRLGDFLNQE SFDLALLEEV WSEQDFQYLR QKLSPTYPAA HHFRSGIIGS GLCVFSKHPI QELTQHIYTL NGYPYMIHHG DWFSGKAVGL LVLHLSGMVL NAYVTHLHAE YNRQKDIYLA HRVAQAWELA QFIHHTSKKA DVVLLCGDLN MHPEDLGCCL LKEWTGLHDA YLETRDFKGS EEGNTMVPKN CYVSQQELKP FPFGVRIDYV LYKAVSGFYI SCKSFETTTG FDPHSGTPLS DHEALMATLF VRHSPPQQNP SSTHGPAERS PLMCVLKEAW TELGLGMAQA RWWATFASYV IGLGLLLLAL LCVLAAGGGA GEAAILLWTP SVGLVLWAGA FYLFHVQEVN GLYRAQAELQ HVLGRAREAQ DLGPEPQPAL LLGQQEGDRT KEQ
Converts sphingomyelin to ceramide. Hydrolyze 1-acyl-2-lyso-sn-glycero-3-phosphocholine (lyso-PC) and 1-O-alkyl-2-lyso-sn-glycero-3-phosphocholine (lyso-platelet-activating factor). The physiological substrate seems to be Lyso-PAF.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lyso-PAF-PLC; Lyso-platelet-activating factor-phospholipase C; N-SMase; Neutral sphingomyelinase; NSMA; Sphingomyelin phosphodiesterase 2; sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase)
Gene Aliases: ISC1; NSMASE; NSMASE1; SMPD2
UniProt ID: (Human) O60906
Entrez Gene ID: (Human) 6610
Molecular Function:
hydrolase
phosphodiesterase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support