Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRLLAWLIFL ANWGGARAEP GKFWHIADLH LDPDYKVSKD PFQVCPSAGS QPVPDAGPWG DYLCDSPWAL INSSIYAMKE IEPEPDFILW TGDDTPHVPD EKLGEAAVLE IVERLTKLIR EVFPDTKVYA ALGNHDFHPK NQFPAGSNNI YNQIAELWKP WLSNESIALF KKGAFYCEKL PGPSGAGRIV VLNTNLYYTS NALTADMADP GQQFQWLEDV LTDASKAGDM VYIVGHVPPG FFEKTQNKAW FREGFNEKYL KVVRKHHRVI AGQFFGHHHT DSFRMLYDDA GVPISAMFIT PGVTPWKTTL PGVVNGANNP AIRVFEYDRA TLSLKVRSPA EARGGGWEGL KCITTFPHSQ LIHLPLTTEP QEG
Located on chromosome 1, this gene encodes for acid sphingomyelinase-like phosphodiesterase 3b precursor protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acid sphingomyelinase-like phosphodiesterase 3b; ASM-like phosphodiesterase 3b; sphingomyelin phosphodiesterase acid-like 3B; sphingomyelin phosphodiesterase, acid-like 3B
Gene Aliases: ASML3B; ASMLPD; SMPDL3B
UniProt ID: (Human) Q92485
Entrez Gene ID: (Human) 27293
Molecular Function:
hydrolase
phosphodiesterase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support