Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: FFGHHHTDSF RMLYDDAGVP ISAMFITPGV TPWKTTLPGV VNGANNPAIR VFEYDRATLS LKVRSPAEAR GGGWEGLKCI TTFPHSQLIH LPLTTEPQE
Located on chromosome 1, this gene encodes for acid sphingomyelinase-like phosphodiesterase 3b precursor protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acid sphingomyelinase-like phosphodiesterase 3b; ASM-like phosphodiesterase 3b; sphingomyelin phosphodiesterase acid-like 3B; sphingomyelin phosphodiesterase, acid-like 3B
Gene Aliases: ASML3B; ASMLPD; SMPDL3B
UniProt ID: (Human) Q92485
Entrez Gene ID: (Human) 27293
Molecular Function:
hydrolase
phosphodiesterase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support