Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Purity is > 95% by SDS-PAGE.
SET domain-containing proteins, such as SMYD2, catalyze lysine methylation..
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Histone methyltransferase SMYD2; HSKM-B; Lysine N-methyltransferase 3C; N-lysine methyltransferase SMYD2; SET and MYND domain-containing protein 2; zinc finger, MYND domain containing 14
Gene Aliases: 1110020E07Rik; 4930402C15; HSKM-B; KMT3C; SMYD2; ZMYND14
UniProt ID: (Human) Q9NRG4, (Mouse) Q8R5A0
Entrez Gene ID: (Human) 56950, (Mouse) 226830
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support