Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Retinoic acid (RA) represents the oxidized form of vitamin A and, via interactions with retinoic acid receptors (RARs), plays a crucial role in development, cellular growth and differentiation. The gene encoding RAI15 maps to human chromosome 2, which houses over 1,400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes. Additionally, an extremely rare recessive genetic disorder, Alström syndrome, is caused by mutations in the ALMS1 gene, which maps to chromosome 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-lysine20 N-trimethyltransferase SMYD5; [histone H4]-lysine36 N-trimethyltransferase SMYD5; Protein NN8 4AG; Protein NN8-4AG; Protein-lysine N-trimethyltransferase SMYD5; retinoic acid induced 15; retinoic acid responsive; retinoic acid responsive gene 1; Retinoic acid-induced protein 15; SET and MYND domain-containing protein 5
Gene Aliases: AW536703; NN8-4AG; RAI15; RRG1; SMYD5; ZMYND23
UniProt ID: (Human) Q6GMV2, (Mouse) Q3TYX3
Entrez Gene ID: (Human) 10322, (Mouse) 232187
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support