Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 8
PA1-738 detects synaptosomal-associated protein 23 (SNAP-23) from human, mouse and rat samples.
PA1-738 has been successfully used in Western blot and immunoprecipitation procedures. By Western blot, this antibody detects an ~23 kDa protein representing SNAP-23 from rat/mouse brain protein extract and HeLa cell lysates.
PA1-738 immunizing peptide corresponds to amino acid residues 193-210 from mouse SNAP-23 protein. PA1-738 immunizing peptide (Cat. # PEP-147) is available for use in neutralization and control experiments.
Specificity of vesicular transport is regulated, in part, by the interaction of a vesicle-associated membrane protein termed synaptobrevin/VAMP with a target compartment membrane protein termed syntaxin. These proteins, together with SNAP25 (synaptosome-associated protein of 25 kDa), form a complex which serves as a binding site for the general membrane fusion machinery. Synaptobrevin/VAMP and syntaxin are believed to be involved in vesicular transport in most, if not all cells, while SNAP25 is present almost exclusively in the brain, suggesting that a ubiquitously expressed homolog of SNAP25 exists to facilitate transport vesicle/target membrane fusion in other tissues. The protein encoded by this gene is structurally and functionally similar to SNAP25 and binds tightly to multiple syntaxins and synaptobrevins/VAMPs. It is an essential component of the high affinity receptor for the general membrane fusion machinery and is an important regulator of transport vesicle docking and fusion. Two alternative transcript variants encoding different protein isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OTTHUMP00000161263; SNAP-23; Synaptosomal-associated protein 23; synaptosomal-associated protein 23 kD; synaptosomal-associated protein, 23 kD; synaptosomal-associated protein, 23kD; synaptosomal-associated protein, 23kDa; synaptosome associated protein 23kDa; Syndet; Vesicle-membrane fusion protein SNAP-23
Gene Aliases: 23kDa; AA408749; HsT17016; SNAP-23; SNAP23; SNAP23A; SNAP23B; Sndt; Syndet
UniProt ID: (Human) O00161, (Mouse) O09044, (Rat) O70377
Entrez Gene ID: (Human) 8773, (Mouse) 20619, (Rat) 64630
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support