Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat and mouse.
Specificity of this antibody: SNIP.
Acts as a negative regulator of SRC by activating CSK which inhibits SRC activity and downstream signaling, leading to impaired cell spreading and migration. Regulates dendritic spine morphology. Involved in calcium-dependent exocytosis. May play a role in neurotransmitter release or synapse maintenance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: P130Cas-associated protein; p140Cap; SNAP-25-interacting protein; SNAP25-interacting protein; SNIP; SRC kinase signaling inhibitor 1
Gene Aliases: KIAA1684; mKIAA1684; P140; p140Cap; SNIP; SRCIN1
UniProt ID: (Human) Q9C0H9, (Mouse) Q9QWI6, (Rat) Q9QXY2
Entrez Gene ID: (Human) 80725, (Mouse) 56013, (Rat) 56029
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support