Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Smad-binding peptide aptamers can be developed to selectively inhibit TGF-beta-induced gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ423B22.2; FHA domain-containing protein SNIP1; RP3-423B22.3; Smad nuclear interacting protein 1; Smad nuclear interacting protein 1 Snip1; Smad nuclear-interacting protein 1
Gene Aliases: 2410133M08Rik; PMRED; SNIP1
UniProt ID: (Human) Q8TAD8, (Rat) Q5M9G6, (Mouse) Q8BIZ6
Entrez Gene ID: (Human) 79753, (Rat) 313588, (Mouse) 76793
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support