Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAGFKRGYDG KIAGLYDLDK TLGRGHFAVV KLARHVFTGE KVAVKVIDKT KLDTLATGHL FQEVRCMKLV QHPNIVRLYE VIDTQTKLYL ILELGDGGDM FDYIMKHEEG LNEDLAKKYF AQIVHAISYC HKLHVVHRDL KPENVVFFEK QGLVKLTDFG FSNKFQPGKK LTTSCGSLAY SAPEILLGDE YDAPAVDIWS LGVILFMLVC GQPPFQEAND SETLTMIMDC KYTVPSHVSK ECKDLITRML QRDPKRRAYL EEIENHPWLQ GVDPSPATKY NIPLVSYKNL SEEEHNSIIQ RMVLGDIADR DAIVEALETN RYNHITATYF LLAERILREK QEKEIQTRSA SPSNIKAQFR QSWPTKIDVP QDLEDDLTAT PLSHATVPQS PARAADSVLN GHRSKGLCDS AKKDDLPELA GPALSTVPPA SLKPTASGRK CLFRVEEDEE EDEEDKKPMS LSTQVVLRRK PSVTNRLTSR KSAPVVNQIF EEGESDDEFD MDENLPPKLS RLKMNIASPG TVHKRYHRRK SQGRGSSCSS SETSDDDSES RRRLDKDSGF TYSWHRRDSS EGPPGSEGDG GGQSKPSNAS GGVDKASPSE NNAGGGSPSS GSGGNPTNTS GTTRRCAGPS NSMQLASRSA GELVESLKLM SLCLGSQLHG STKYIIDPQN GLSFSSVKVQ EKSTWKMCIS STGNAGQVPA VGGIKFFSDH MADTTTELER IRSKNLKNNV LQLPLCEKTI SVNIQRNPKE GLLCASSPAS CCHVI
SNRK may play a role in hematopoietic cell proliferation or differentiation. SNRK is the potential mediator of neuronal apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SNF-1 related kinase; SNF-related serine/threonine-protein kinase; SNF1-related kinase; SNRK
Gene Aliases: HSNFRK; KIAA0096; SNFRK; SNRK
UniProt ID: (Human) Q9NRH2
Entrez Gene ID: (Human) 54861
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support