Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total SNRK.
SNRK may play a role in hematopoietic cell proliferation or differentiation. SNRK is the potential mediator of neuronal apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SNF-1 related kinase; SNF-related serine/threonine-protein kinase; SNF1-related kinase; SNRK; snrk {ECO:0000312|RGD:69653}
Gene Aliases: 2010012F07Rik; AI448042; AW547029; E030034B15; HSNFRK; KIAA0096; mKIAA0096; R74830; SNFRK; SNRK
UniProt ID: (Human) Q9NRH2, (Mouse) Q8VDU5, (Rat) Q63553
Entrez Gene ID: (Human) 54861, (Mouse) 20623, (Rat) 170837
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support