Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ELVEQTLSD LEQSKCISIE DEMDVAPLNL GMIAAYYYIN YTTIELFSMS LNAKTKVRGL IEIISNAAEY ENIPIRHHED NLLRQLAQKV PHKLNNPKFN DPHVKTNLLL QAHLSRMQLS AELQSDTEEI LSKAIRLIQA CVDVLSSNGW LSPALAAMEL AQMVTQAMWS KDSYLKQLPH FTSEHIKRCT DKGVESVFDI MEMEDEERNA LLQLTDSQIA DVARFCNRYP NIELSYEVVD KDSIRSGGPV VVLVQLEREE EVTGPVIAPL FPQKREEGWW VVIGDAKSNS LISIKRLTLQ QKAKVKLDFV APATGAHNYT LYFMSDAYMG CDQEYKFSVD VKEAETDSDS D (1787-2136 aa encoded by BC001417)
Pre-mRNA splicing is catalyzed by the spliceosome, a complex of specialized RNA and protein subunits that removes introns from a transcribed pre-mRNA segment. The spliceosome consists of small nuclear RNA proteins (snRNPs) U1, U2, U4, U5 and U6, together with approximately 80 conserved proteins. U5 snRNP contains nine specific proteins. This gene encodes one of the U5 snRNP-specific proteins. This protein belongs to the DEXH-box family of putative RNA helicases. It is a core component of U4/U6-U5 snRNPs and appears to catalyze an ATP-dependent unwinding of U4/U6 RNA duplices. Mutations in this gene cause autosomal-dominant retinitis pigmentosa. Alternatively spliced transcript variants encoding different isoforms have been found, but the full-length nature of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating signal cointegrator 1 complex subunit 3-like 1; bad response to refrigeration 2 homolog; BRR2 homolog; small nuclear ribonucleoprotein 200kDa (U5); small nuclear ribonucleoprotein, U5 200kDa subunit; U5 small nuclear ribonucleoprotein 200 kDa helicase; U5 snRNP-specific 200 kDa protein; U5 snRNP-specific protein, 200 kDa (DEXH RNA helicase family); U5-200KD
Gene Aliases: A330064G03Rik; ASCC3L1; BC011390; BRR2; HELIC2; KIAA0788; RP33; SNRNP200; U5-200-KD; U5-200KD
UniProt ID: (Human) O75643, (Mouse) Q6P4T2
Entrez Gene ID: (Human) 23020, (Mouse) 320632
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support